Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MAP2K4 (MEK4), GST Tag Recombinant Protein

Catalog No. A32894
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 μg
100 μg

Recombinant Protein

MAP2K4 (MEK4) serine/threonine kinase (mitogen-activated protein kinase 4) MEK4 or MKK4 is a dual specificity kinase of the STE7 family that phosphorylates and activates JNK1 and -2 as well as p38. MAP2K4 preferentially phosphorylates the tyrosine residue of JNKs. MAP2K4 mediates a variety of physiological and pathophysiological processes such as responses to cellular stresses and inflammatory cytokines. MAP2K4 is important for carcinogenesis; a loss-off-function mutation in MAP2K4 was found in lung and pancreatic tumors. Conversely, a pro-oncogenic role was also found for MAP2K4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P45985
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 6416
Molecular Weight (g/mol) 70.6 kDa
Name Human MAP2K4 (MEK4), GST Tag
Quantity 100 μg
Gene Alias C-JUN N-terminal kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 2; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4; dual specificity mitogen-activated protein kinase kinase 4 A; JNK kinase 1; JNK-activated kinase 1; JNK-activating kinase 1; JNKK; JNKK 1; JNKK1; MAP kinase activator 2; MAP kinase kinase 4; Map2k4; map2k4.L; map2k4a; map2k4-a; map2k4-b; MAP2K4delta; MAPK/ERK kinase 4; MAPKK 4; MAPKK4; MEK 4; MEK4; mitogen activated protein kinase kinase 4; mitogen activated protein kinase kinase 4, variant 1; mitogen-activated protein kinase kinase 4; mitogen-activated protein kinase kinase 4 L homeolog; mitogen-activated protein kinase kinase 4 A; MKK4; mkk4a; MKK4A-l; PRKMK4; SAPK kinase 1; SAPK/ERK kinase 1; SAPK/Erk/kinase 1; SAPKK1; SAPKK-1; Sek1; Serk1; Skk1; Stress-activated protein kinase kinase 1; XELAEV_18044157mg; XMEK2; zgc:136500; zMKK4 protein
Common Name MAP2K4
KinaseFamily MAP Kinases
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Full Length, Recombinant, Active
Form Liquid
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.