Learn More
MAP2K4 Mouse anti-Canine, Human, Clone: OTI8A8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500403

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MAP2K4 (SEK1) is a dual specificity protein kinase that belongs to the Ser/Thr protein kinase family. This kinase is a direct activator of MAP kinases in response to various environmental stresses or mitogenic stimuli. It has been shown to activate MAPK8/JNK1, MAPK9/JNK2, and MAPK14/p38, but not MAPK1/ERK2 or MAPK3/ERK3. MAP2K4 is phosphorylated and activated by MAP3K1/MEK.Specifications
MAP2K4 | |
Monoclonal | |
Unconjugated | |
MAP2K4 | |
JNKK, JNKK1, MAPKK4, MEK4, MKK4, PRKMK4, SAPKK-1, SAPKK1, SEK1, SERK1, SKK1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
489508, 6416 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
OTI8A8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P45985 | |
MAP2K4 | |
Full-length protein expressed in 293T cell transfected with human MAP2K4 expression vector | |
100 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.