Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MAP2K5 Recombinant Protein

Catalog No. p-3720016
Click to view available options
Quantity:
10 μg
25 μg

Recombinant protein for MAP2K5 (human) gene

The encoded protein is a dual specificity protein kinase that belongs to the MAP kinase kinase family. This kinase specifically interacts with and activates MAPK7/ERK5. This kinase itself can be phosphorylated and activated by MAP3K3/MEKK3, as well as by atypical protein kinase C isoforms (aPKCs).

  • Human MAP2K5 full-length ORF ( AAH08838, 1 a.a. - 448 a.a.) recombinant protein with GST-tag at N-terminal
  • Description: mitogen-activated protein kinase 5
  • Theoretical molecular weight: 75.02kDa
  • Preparation: in vitro wheat germ expression system
  • Purification: glutathione sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced glutathione, pH: 8.0 in elution buffer

Sequence: MLWLALGPFPAMENQVLVIRIKIPNSGAVDWTVHSGPQLLFRDVLDVIGQVLPEATTTAFEYEDEDGDRITVRSDEEMKAMLSYYYSTVMEQQVNGQLIEPLQIFPRACKPPGERNIHGLKVNTRAGPSQHSSPAVSDSLPSNSLKKSSAELKKILANGQMNEQDIRYRDTLGHGNGGTVYKAYH VPSGKILAVKVILLDITLELQKQIMSELEILYKCDSSYIIGFYGAFFVENRISICTEFMDGGSLDVYRKMPEHVLGRIAVAVVKGLTYLWSLKILHRDVKPSNMLVNTRGQVKLCDFGVSTQLVNSIAKTYVGTNAYMAPERISGEQYGIHSDVWSLGISFMELALGRFPYPQIQKNQGSLMPLQLLQCIVDEDSPVLPVGESSEPFVHFITQCMRKQPKERPAPEELMGHPFIVQFNDGNAAVVSMWVCRALEERRSQQGPP

Best use within three months from the date of receipt.

ELISA, western blotting (recombinant protein), antibody production, protein array

Specifications

Accession Number AAH08838
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 5607
Molecular Weight (g/mol) 75.02
Name MAP2K5 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MLWLALGPFPAMENQVLVIRIKIPNSGAVDWTVHSGPQLLFRDVLDVIGQVLPEATTTAFEYEDEDGDRITVRSDEEMKAMLSYYYSTVMEQQVNGQLIEPLQIFPRACKPPGERNIHGLKVNTRAGPSQHSSPAVSDSLPSNSLKKSSAELKKILANGQMNEQDIRYRDTLGHGNGGTVYKAYHVPSGKILAVKVILLDITLELQKQIMSELEILYKCDSSYIIGFYGAFFVENRISICTEFMDGGSLDVYRKMPEHVLGRIAVAVVKGLTYLWSLKILHRDVKPSNMLVNTRGQVKLCDFGVSTQLVNSIAKTYVGTNAYMAPERISGEQYGIHSDVWSLGISFMELALGRFPYPQIQKNQGSLMPLQLLQCIVDEDSPVLPVGESSEPFVHFITQCMRKQPKERPAPEELMGHPFIVQFNDGNAAVVSMWVCRALEERRSQQGPP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias HsT17454/MAPKK5/MEK5/PRKMK5
Common Name MAP2K5
Gene Symbol MAP2K5
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.