Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEKK1 Monoclonal Antibody (EBS-T-008)
GREENER_CHOICE

Catalog No. PIMA533332
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533332 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533332 Supplier Invitrogen™ Supplier No. MA533332
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MAPKs are involved in directing cellular responses to a diverse array of potentially harmful stimuli, such as mitogens, osmotic stress, heat shock, proinflammatory cytokines, but also growth factors (mammals). Possibly located exclusively in the cell nucleus, they regulate cell functions including proliferation, gene expression, differentiation, mitosis, cell survival, and apoptosis. In order to become active, they require usually multiple phosphorylation events in their activation loops, including phosphorylation by MAP2 kinases (Ste-7 kinases), which in turn are phosphorylated by the MAP3 kinase family, of which many are located at the cell membrane. Thus through this pathway, stimuli can effectively be conveyed from the cell membrane to the nucleus. Inactivation of MAPKs takes place by several phosphorylases, including dedicated phophorylases.

Mitogen-activated protein kinase (MAPK) signaling cascades include MAPK or extracellular signal-regulated kinase (ERK), MAPK kinase (MKK or MEK), and MAPK kinase kinase (MAPKKK or MEKK). MAPKK kinase/MEKK phosphorylates and activates its downstream protein kinase, MAPK kinase/MEK, which in turn activates MAPK. The kinases of these signaling cascades are highly conserved, and homologs exist in yeast, Drosophila, and mammalian cells. MEKK1 can phosphorylate and activate MAPKK 1 and MAPKK 2 (MEK1/MEK2) which leads to phosphorylation of MAP kinases. It is also a highly efficient activator of the JNK cascade. The protein contains a putative 1 RING-type zinc finger and 1 SWIM-type zinc finger.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEKK1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone EBS-T-008
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MAP3K1
Gene Accession No. Q13233
Gene Alias MAP/ERK kinase kinase 1; MAP3K1; MAPK/ERK kinase kinase 1; MAPKKK1; MEK kinase 1; MEKK; MEKK 1; Mekk1; mitogen activated protein kinase kinase kinase 1; mitogen-activated protein kinase kinase kinase 1; mitogen-activated protein kinase kinase kinase 1, E3 ubiquitin protein ligase; si:rp71-80a19.2; SRXY6
Gene Symbols MAP3K1
Host Species Mouse
Immunogen Partial recombinant MAP3K1 (aa1211-1310).
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4214
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.