Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP3K12 binding inhibitory protein 1 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Specifications
| Antigen | MAP3K12 binding inhibitory protein 1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
MAP3K12 binding inhibitory protein 1 Polyclonal specifically detects MAP3K12 binding inhibitory protein 1 in Rat samples. It is validated for Western Blot.Specifications
| MAP3K12 binding inhibitory protein 1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| MAP3K12 binding inhibitory protein 1, MAP3K12-binding inhibitory protein 1, MAPK upstream kinase-binding inhibitory protein, MUK-binding inhibitory protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-344 of human MAP3K12 binding inhibitory protein 1 (NP_057670.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS with 50% glycerol, pH7.3. | |
| 51562 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title