Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP3K12 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc MAP3K12101AP

Description
MAP3K12 Polyclonal antibody specifically detects MAP3K12 in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western BlotSpecifications
MAP3K12 | |
Polyclonal | |
Unconjugated | |
MAP3K12 | |
DLK; Dual leucine zipper bearing kinase; dual leucine zipper kinase; dual leucine zipper kinase DLK; leucine zipper protein kinase; leucine-zipper protein kinase; M3KC; MAP3K12; MAPK-upstream kinase; MEKK12; Mitogen activated protein kinase 12 (Zipper (leucine) protein kinase); mitogen activated protein kinase kinase kinase 12; mitogen-activated protein kinase kinase kinase 12; Mixed lineage kinase; MUK; PK; protein kinase MUK; zipper (leucine) protein kinase; Zpk; ZPKP1 | |
Synthetic peptide taken within amino acid region 270-320 on MAP3K12 protein | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
Q12852, Q60700, Q63796 | |
Rabbit | |
Affinity chromatography | |
RUO | |
25579, 26404, 7786 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction