Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP3K15 Antibody (OTI1G10), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272588B
Description
MAP3K15 Monoclonal specifically detects MAP3K15 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MAP3K15 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
MAP3K15 | |
Recombinant protein fragment corresponding to amino acids 979-1313 of human MAP3K15 (NP_001001671) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI1G10 | |
Western Blot | |
Apoptosis signal-regulating kinase 3, ASK3MEKK 15, bA723P2.3, EC 2.7.11, EC 2.7.11.25, FLJ16518, MAPK/ERK kinase kinase 15, MEK kinase 15, mitogen-activated protein kinase kinase kinase 15 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
389840.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction