Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP3K15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103334
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103334 Supplier Invitrogen™ Supplier No. PA5103334
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MAP3K15.

MAP3K15 is a component of a protein kinase signal transduction cascade.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP3K15
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MAP3K15
Gene Accession No. A2AQW0, Q6ZN16
Gene Alias apoptosis signal-regulating kinase 3; ASK3; bA723P2.3; BC031147; map3k15; MAPK/ERK kinase kinase 15; MEK kinase 15; MEKK 15; MEKK15; mitogen-activated protein kinase kinase kinase 15
Gene Symbols MAP3K15
Host Species Rabbit
Immunogen A synthesized peptide derived from human MAP3K15(Accession Q6ZN16), corresponding to amino acid residues E1210-K1260.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 270672, 389840
Target Species Human, Mouse, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.