Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAP3K6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119289
This item is not returnable.
View return policy
Description
MAP3K6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Mitogen-activated protein kinase (MAPK) signaling cascades include MAPK or extracellular signal-regulated kinase (ERK), MAPK kinase (MKK or MEK), and MAPK kinase kinase (MAPKKK or MEKK). MAPKK kinase/MEKK phosphorylates and activates its downstream protein kinase, MAPK kinase/MEK, which in turn activates MAPK. The kinases of these signaling cascades are highly conserved, and homologs exist in yeast, Drosophila, and mammalian cells. MEKK6 activates the JNK, but not ERK or p38 kinase pathways.
Specifications
MAP3K6 | |
Polyclonal | |
Unconjugated | |
MAP3K6 | |
apoptosis signal-regulating kinase 2; ASK2; map3k6; MAPKKK6; MEKK6; MGC125653; MGC20114; mitogen activated protein kinase kinase kinase 6; mitogen-activated protein kinase kinase kinase 6; OTTHUMP00000005063 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
313022 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
0 | |
MAP3K6 | |
Recombinant protein Mitogen Activated Protein Kinase Kinase Kinase 6. | |
200 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction