Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP3K8/Tpl2/COT Antibody (HL2118) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325556
Description
MAP3K8/Tpl2/COT Monoclonal antibody specifically detects MAP3K8/Tpl2/COT in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
MAP3K8/Tpl2/COT | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HL2118 | |
Western Blot 1:500-1:3000, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Cancer Osaka thyroid oncogene, c-COT, cot (cancer Osaka thyroid) oncogene, COTMEKK8, EC 2.7.11.25, ESTF, ESTFLJ10486, Ewing sarcoma transformant, mitogen-activated protein kinase kinase kinase 8, Proto-oncogene c-Cot, proto-oncogene serine/threoine protein kinase, Serine/threonine-protein kinase cot, Tpl-2, TPL2, Tumor progression locus 2, tumor progression locus-2 | |
Recombinant fragmemt of human MAP3K8/Tpl2/COT. | |
100 μL | |
Angiogenesis, Cancer, Protein Kinase | |
1326 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction