Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP3K8/Tpl2/COT Antibody (OTI3D9), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272589AF405
Description
MAP3K8/Tpl2/COT Monoclonal specifically detects MAP3K8/Tpl2/COT in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MAP3K8/Tpl2/COT | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAP3K8 | |
Recombinant protein fragment corresponding to amino acids 277-461 of human MAP3K8/Tpl2/COT (NP_005195) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI3D9 | |
Western Blot | |
Cancer Osaka thyroid oncogene, c-COT, cot (cancer Osaka thyroid) oncogene, COTMEKK8, EC 2.7.11.25, ESTF, ESTFLJ10486, Ewing sarcoma transformant, mitogen-activated protein kinase kinase kinase 8, Proto-oncogene c-Cot, proto-oncogene serine/threoine protein kinase, Serine/threonine-protein kinase cot, Tpl-2, TPL2, Tumor progression locus 2, tumor progression locus-2 | |
Mouse | |
Affinity Purified | |
Angiogenesis, Cancer, Protein Kinase | |
1326.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction