Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP3K9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18278125UL
Description
MAP3K9 Polyclonal specifically detects MAP3K9 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MAP3K9 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| EC 2.7.11, EC 2.7.11.25, MEKK9, mitogen-activated protein kinase kinase kinase 9, MLK1Mixed lineage kinase 1, PRKE1mixed lineage kinase 1 (tyr and ser/thr specificity) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| MAP3K9 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:EGSPQRREKANGLSTPSESPHFHLGLKSLVDGYKQWSSSAPNLVKGPRSSPALPGFTSLMEMEDEDSEGPGSGESRLQHSPSQSYLCIPFPRGEDGDGPSSDGIHEEPTPVNSATSTPQLTPTNSLKRGG | |
| 25 μL | |
| GPCR, MAP Kinase Signaling, Protein Kinase | |
| 4293 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction