Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558272 Supplier Invitrogen™ Supplier No. PA558272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAPLAKDGVL TLANNVTPAK DVPPLSETEA TPVPIKDMEI AQTQKGISED SHLESLQDVG QSAAPTFMIS PETVTGT Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 45%.

The protein encoded by this gene is a major non-neuronal microtubule-associated protein. This protein contains a domain similar to the microtubule-binding domains of neuronal microtubule-associated protein (MAP2) and microtubule-associated protein tau (MAPT/TAU). This protein promotes microtubule assembly, and has been shown to counteract destabilization of interphase microtubule catastrophe promotion. Cyclin B was found to interact with this protein, which targets cell division cycle 2 (CDC2) kinase to microtubules. The phosphorylation of this protein affects microtubule properties and cell cycle progression. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAP4
Gene Accession No. P27816
Gene Alias AA407148; MAP 4; Map4; MAP-4; microtubule associated protein 4; microtubule-associated protein 4; Mtap4; Mtap-4
Gene Symbols MAP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAP4. Recombinant protein control fragment (Product #RP-96264).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4134
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.