Learn More
Invitrogen™ MAP4 Polyclonal Antibody

Description
Immunogen sequence: TENIKHQPGG GRAKVEKKTE AAATTRKPES NAVTKTAGPI ASAQKQPAGK VQIVSKKVSY SHIQSKCGSK DNIKHVPGGG NVQIQNKKVD ISKVSSKCGS KANIKHKPGG GDVKIESQKL NFKEKAQAKV GSLDNVGHLP AGGAVKTEGG GSEAPLCPGP PAGEEPAISE AAPEAGAPTS ASGLNGHPTL SGGGDQREAQ TLDSQIQETS I; Positive Samples: U-87MG; Cellular Location: Cytoplasm, cytoskeleton.
Specifications
Specifications
| Antigen | MAP4 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.09 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | MAP4 |
| Gene Accession No. | P27546, P27816 |
| Gene Alias | AA407148; MAP 4; Map4; MAP-4; microtubule associated protein 4; microtubule-associated protein 4; Mtap4; Mtap-4 |
| Gene Symbols | MAP4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.