Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP4K2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP188584
Description
MAP4K2 Polyclonal specifically detects MAP4K2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MAP4K2 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
B lymphocyte serine/threonine protein kinase, B lymphocyte serine/threonine-protein kinase, BL44, EC 2.7.11, EC 2.7.11.1, GC kinase, GCKMEK kinase kinase 2, Germinal center kinase, germinal centre kinase (GC kinase), MAPK/ERK kinase kinase kinase 2, mitogen-activated protein kinase kinase kinase kinase 2, Rab8 interacting protein, Rab8-interacting protein, RAB8IPMEKKK 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
MAP4K2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ETDPLNEPWEEEWTLLGKEELSGSLLQSVQEALEERSLTIRSASEFQELDSPDDTMGTIKRAPFLGPLPTDPPAEEPLSSPPGTLPPPPSGPNSSPLLPTAWATMKQRE | |
0.1 mL | |
Signal Transduction | |
5871 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction