Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP4K2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552422
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552422 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552422 Supplier Invitrogen™ Supplier No. PA552422
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ETDPLNEPWE EEWTLLGKEE LSGSLLQSVQ EALEERSLTI RSASEFQELD SPDDTMGTIK RAPFLGPLPT DPPAEEPLSS PPGTLPPPPS GPNSSPLLPT AWATMKQRE Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 74%.

MAP4K2 (GCK) is a serine/ threonine protein kinase that activates the JNK pathway. Although this kinase is found in many tissues, its expression in lymphoid follicles is restricted to the cells of the germinal center, where it may participate in B-cell differentiation. The kinase may also play a role in the regulation of vesicle targeting or fusion. The protein encoded by MAP4K2 is a member of the serine/threonine protein kinase family. Although this kinase is found in many tissues, its expression in lymphoid follicles is restricted to the cells of germinal centre, where it may participate in B-cell differentiation. This kinase can be activated by TNF-alpha, and has been shown to specifically activate MAP kinases. This kinase is also found to interact with TNF receptor-associated factor 2 (TRAF2), which is involved in the activation of MAP3K1/MEKK1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP4K2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAP4K2
Gene Accession No. Q12851
Gene Alias AI385662; B lymphocyte serine/threonine protein kinase; B lymphocyte serine/threonine-protein kinase; BL44; dZ150L22.2; GC kinase; GCK; germinal center kinase; germinal centre kinase (GC kinase); LOW QUALITY PROTEIN: mitogen-activated protein kinase kinase kinase kinase 2; MAP4K2; map4k2l; map4k2l-2; map4k3l; MAPK/ERK kinase kinase kinase 2; MEK kinase kinase 2; MEKKK 2; mitogen activated protein kinase kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase kinase 2-like; mitogen-activated protein kinase kinase kinase kinase 3-like; Rab8 interacting protein; rab8-interacting protein; Rab8ip; si:dz150l22.2; si:dz263j20.1; Unknown (protein for MGC:136670); zgc:136670
Gene Symbols MAP4K2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAP4K2. Recombinant protein control fragment (Product #RP-89768).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5871
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.