Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAP4K2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116226
Description
MAP4K2 Polyclonal Antibody for Western Blot
MAP4K2 (GCK) is a serine/ threonine protein kinase that activates the JNK pathway. Although this kinase is found in many tissues, its expression in lymphoid follicles is restricted to the cells of the germinal center, where it may participate in B-cell differentiation. The kinase may also play a role in the regulation of vesicle targeting or fusion. The protein encoded by MAP4K2 is a member of the serine/threonine protein kinase family. Although this kinase is found in many tissues, its expression in lymphoid follicles is restricted to the cells of germinal centre, where it may participate in B-cell differentiation. This kinase can be activated by TNF-alpha, and has been shown to specifically activate MAP kinases. This kinase is also found to interact with TNF receptor-associated factor 2 (TRAF2), which is involved in the activation of MAP3K1/MEKK1.
Specifications
MAP4K2 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q61161 | |
MAP4K2 | |
KLH conjugated synthetic peptide between 432-459 amino acids from the Central region of mouse Map4k2. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MAP4K2 | |
AI385662; B lymphocyte serine/threonine protein kinase; B lymphocyte serine/threonine-protein kinase; BL44; dZ150L22.2; GC kinase; GCK; germinal center kinase; germinal centre kinase (GC kinase); LOW QUALITY PROTEIN: mitogen-activated protein kinase kinase kinase kinase 2; MAP4K2; map4k2l; map4k2l-2; map4k3l; MAPK/ERK kinase kinase kinase 2; MEK kinase kinase 2; MEKKK 2; mitogen activated protein kinase kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase kinase 2-like; mitogen-activated protein kinase kinase kinase kinase 3-like; Rab8 interacting protein; rab8-interacting protein; Rab8ip; si:dz150l22.2; si:dz263j20.1; Unknown (protein for MGC:136670); zgc:136670 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
26412 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction