Learn More
Invitrogen™ MAP4K3 Polyclonal Antibody

Description
MAP4K3 Polyclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | MAP4K3 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | MAP4K3 |
| Gene Accession No. | Q8IVH8 |
| Gene Alias | 4833416M01Rik; 4833416M07Rik; 9530052P13Rik; germinal center kinase-like kinase; germinal center kinase-related protein kinase; GLK; MAP4K3; MAPK/ERK kinase kinase kinase 3; MAPKKKK3; MEK kinase kinase 3; MEKKK 3; MEKKK3; mitogen activated protein kinase kinase kinase kinase 3; mitogen-activated protein kinase kinase kinase kinase 3; RAB8IPL1 |
| Gene Symbols | MAP4K3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.