Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP4K4/HGK Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

$151.31

Specifications

Antigen MAP4K4/HGK
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-156-0556
Encompass_Preferred
View Documents
Bethyl Laboratories, Inc
A301502AT
20 μL
Each of 1 for $151.31
Only null left
Add to Cart
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

M4K4, a member of the STE20 subfamily of Ser/Thr protein kinases, may play a role in the response to environmental stress and cytokines such as TNF-alpha. It appears to act upstream of the JUN N-terminal pathway. This protein is thought to interact with the SH3 domain of the adapter proteins Nck. HGK binds, via its CNH regulatory domain, to the N-terminal region of SPG3A. Expression appears to be ubiquitous, expressed in all tissue types examined. Isoform 5 appears to be more abundant in the brain, and isoform 4 is predomit in the liver, skeletal muscle and placenta.
Specifications

Specifications

MAP4K4/HGK
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
Mitogen-activated protein kinase kinase kinase kinase 4; MAPK/ERK kinase kinase kinase 4; MEK kinase kinase 4; HPK/GCK-like kinase HGK; Nck interacting kinase; MEKKK 4; MAP4K4; NIK; KIAA0687
MAP4K4
IgG
Antigen affinity chromatography
Antibody
Immunohistochemistry, Western Blot
Unconjugated
Rabbit
Human
O95819
9448
Between 550 and 600
Primary
4° C
MAP4K4
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.