Learn More
MAP4K4/HGK Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301503A
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
M4K4, a member of the STE20 subfamily of Ser/Thr protein kinases, may play a role in the response to environmental stress and cytokines such as TNF-alpha. It appears to act upstream of the JUN N-terminal pathway. This protein is thought to interact with the SH3 domain of the adapter proteins Nck. HGK binds, via its CNH regulatory domain, to the N-terminal region of SPG3A. Expression appears to be ubiquitous, expressed in all tissue types examined. Isoform 5 appears to be more abundant in the brain, and isoform 4 is predomit in the liver, skeletal muscle and placenta.Specifications
| MAP4K4/HGK | |
| Polyclonal | |
| Unconjugated | |
| MAP4K4 | |
| Mitogen-activated protein kinase kinase kinase kinase 4; MAPK/ERK kinase kinase kinase 4; MEK kinase kinase 4; HPK/GCK-like kinase HGK; Nck interacting kinase; MEKKK 4; MAP4K4; NIK; KIAA0687 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9448 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| O95819 | |
| MAP4K4 | |
| Between 850 and 900 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.