Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP4K5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557775
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557775 Supplier Invitrogen™ Supplier No. PA557775
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YPEDNFPDEE KASTIKHCPD SESRAPQILR RQSSPSCGPV AETSSIGNGD GISKLMSENT EGSAQAPQLP RKKDKRDFPK PA Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 82%.

MAP4K5 (KHS1) is a serine/threonine kinase that belongs to the GCK family of kinases and has been implicated as an upstream regulator of MAP kinase signaling pathways. Yeast SPS1/STE20 functions near the beginning of the MAP kinase signal cascades that is essential for yeast pheromone response. This kinase was shown to activate Jun kinase in mammalian cells, which suggested a role in stress response. Two alternatively spliced transcript variants encoding the same protein have been described for MAP4K5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP4K5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Map4k5
Gene Accession No. Q9Y4K4
Gene Alias 4432415E19Rik; fl74c10; GCKR; germinal center kinase-related; KHS; KHS1; Kinase homologous to SPS1/STE20; LOW QUALITY PROTEIN: mitogen-activated protein kinase kinase kinase kinase 5; map4k5; MAPK/ERK kinase kinase kinase 5; MAPKKKK5; MEK kinase kinase 5; MEKKK 5; mitogen-activated protein kinase kinase kinase kinase 5; RGD1562028; wu:fl74c10; zgc:55719; zgc:55985
Gene Symbols Map4k5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAP4K5. Recombinant protein control fragment (Product #RP-97238).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11183
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.