Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP4K5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983052
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3052 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3052 Supplier Bioss Supplier No. BS13654R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAP4K5 may play a role in the response to environmental stress. Appears to act upstream of the JUN N-terminal pathway. This gene encodes a member of the serine/threonine protein kinase family, that is highly similar to yeast SPS1/STE20 kinase. Yeast SPS1/STE20 functions near the beginning of the MAP kinase signal cascades that is essential for yeast pheromone response. This kinase was shown to activate Jun kinase in mammalian cells, which suggested a role in stress response. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.

Specifications

Antigen MAP4K5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Map4k5
Gene Accession No. Q9Y4K4
Gene Alias 4432415E19Rik; BOS_10813; fl74c10; GCKR; germinal center kinase-related; KHS; KHS1; Kinase homologous to SPS1/STE20; LOW QUALITY PROTEIN: mitogen-activated protein kinase kinase kinase kinase 5; Map4k5; MAPK/ERK kinase kinase kinase 5; MAPKKKK5; MEK kinase kinase 5; MEKKK 5; Mitogen-activated protein kinase kinase kinase kinase 5; RGD1562028; wu:fl74c10; zgc:55719; zgc:55985
Gene Symbols Map4k5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MAP4K5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11183
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.