Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAP6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558640
Description
Immunogen sequence: TSASHKPTRK AKDKQAVSGQ AAKKKSAEGP STTKPDDKEQ SKEMNNKLAE AKESLAQPVS DSSKTQGPVA TEPDKDQGSV VPGLLKGQGP MVQEPLKKQG SVVPGPPKDL GPM Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.
STOP encodes a microtubule-associated protein. The encoded protein is a calmodulin-binding and calmodulin-regulated protein that is involved in microtubule stabilization. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
MAP6 | |
Polyclonal | |
Unconjugated | |
Map6 | |
145-kDa STOP; 2810411E12Rik; CTD-2530H12.7; KIAA1878; LOW QUALITY PROTEIN: microtubule-associated protein 6; MAP6; MAP-6; MAP6-N; microtubule associated protein 6; microtubule-associated protein 6; microtubule-associated protein 6 homolog; Mtap6; N STOP; N-STOP; Stable tubule-only polypeptide; STOP; STOP145 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4135 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JE9 | |
Map6 | |
Recombinant protein corresponding to Human MAP6. Recombinant protein control fragment (Product #RP-97466). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction