Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578399
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578399 Supplier Invitrogen™ Supplier No. PA578399
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: rat brain, mouse brain Predicted Reactivity: Human (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

STOP encodes a microtubule-associated protein. The encoded protein is a calmodulin-binding and calmodulin-regulated protein that is involved in microtubule stabilization. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP6
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Map6
Gene Accession No. Q63560, Q7TSJ2, Q96JE9
Gene Alias 145-kDa STOP; 2810411E12Rik; CTD-2530H12.7; KIAA1878; LOW QUALITY PROTEIN: microtubule-associated protein 6; MAP6; MAP-6; MAP6-N; microtubule associated protein 6; microtubule-associated protein 6; microtubule-associated protein 6 homolog; Mtap6; N STOP; N-STOP; Stable tubule-only polypeptide; STOP; STOP145
Gene Symbols Map6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human STOP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17760, 29457, 4135
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.