Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545077
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545077 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545077 Supplier Invitrogen™ Supplier No. PA545077
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IRSLYSEPFK EPPKVEKPSV QSSKPKKTSA SHKPTRKAKD KQAVSGQAAK Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 86%.

STOP encodes a microtubule-associated protein. The encoded protein is a calmodulin-binding and calmodulin-regulated protein that is involved in microtubule stabilization. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Map6
Gene Accession No. Q96JE9
Gene Alias 145-kDa STOP; 2810411E12Rik; CTD-2530H12.7; KIAA1878; LOW QUALITY PROTEIN: microtubule-associated protein 6; MAP6; MAP-6; MAP6-N; microtubule associated protein 6; microtubule-associated protein 6; microtubule-associated protein 6 homolog; Mtap6; N STOP; N-STOP; Stable tubule-only polypeptide; STOP; STOP145
Gene Symbols Map6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MAP6.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4135
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.