Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAP6D1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571406
Description
MAP6D1 Polyclonal Antibody for Western Blot
This gene encodes a protein highly similar to the mouse MAP6 domain containing 1 protein, which is related to the STOP proteins. Based on the study of the mouse protein, the encoded protein may function as a calmodulin-regulated neuronal protein that binds and stabilizes microtubules but also associates with the Golgi membranes through N-terminal palmitoylation.
Specifications
MAP6D1 | |
Polyclonal | |
Unconjugated | |
MAP6D1 | |
21 kDa STOP-like protein; MAP6 domain containing 1; MAP6 domain-containing protein 1; MAP6D1; MAPO6D1; SL21; STOP-Like protein 21 kD | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79929 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H9H5 | |
MAP6D1 | |
synthetic peptide directed towards the middle region of human MAP6D1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction