Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556235
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556235 Supplier Invitrogen™ Supplier No. PA556235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PHLPGTPRPT SSLPPGSVKA APAQVRPPSP GNIRPVKREV KVEPEKKDPE KEPQKVANEP SLKGRAPLVK VEEATVEERT PAEPEVGPAA P Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 70%.

The product of this gene is a microtubule-associated protein that is predominantly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAP7
Gene Accession No. Q14244
Gene Alias dJ325F22.2 (microtubule-associated protein 7 (EMAP115, E-MAP-115)); EMAP115; E-MAP-115; Ensconsin; epithelial microtubule-associated protein of 115 kDa; MAP7; MAP-7; microtubule associated protein 7; microtubule-associated protein 7; mshi; mste; Mtap7; R75000; ste
Gene Symbols MAP7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MAP7. Recombinant protein control fragment (Product #RP-94845).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9053
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.