Learn More
MAP7 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301491A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Mouse
The product of this gene is a microtubule-associated protein that is predomitly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis.Specifications
| MAP7 | |
| Polyclonal | |
| Unconjugated | |
| MAP7 | |
| E-MAP-115, EMAP115, MAP-7, dJ325F22.2 (microtubule-associated protein 7 (EMAP115, E-MAP-115)), ensconsin, epithelial microtubule-associated protein of 115 kDa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9053 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q14244 | |
| MAP7 | |
| Between 720 and 749 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.