Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MAP7D1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544693
Description
Peptide sequence: RRRLEEQRLK AEQRRAALEE RQRQKLEKNK ERYEAAIQRS VKKTWAEIRQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
MAP7D1 (MAP7 Domain Containing 1) is a Protein Coding gene.
Specifications
MAP7D1 | |
Polyclonal | |
Unconjugated | |
Map7d1 | |
arginine/proline rich coiled-coil 1; arginine/proline-rich coiled-coil domain-containing protein 1; AV028413; BC019977; FLJ10350; FLJ39022; KIAA1187; MAP7 domain containing 1; MAP7 domain-containing protein 1; MAP7D1; microtubule-associated protein 7 domain containing 1; mKIAA1187; Mtap7d1; Parcc1; PP2464; proline arginine rich coiled coil 1; proline/arginine-rich coiled-coil domain-containing protein 1; Rprc1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
245877, 55700 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A2AJI0, Q3KQU3 | |
Map7d1 | |
Synthetic peptide directed towards the N-terminal of human MAP7D1 (aa 179-228). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction