Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MAP7D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590770
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590770 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590770 Supplier Invitrogen™ Supplier No. PA590770
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QRQKLEKNKE RYEAAIQRSV KKTWAEIRQQ RWSWAGALHH SSPGHKTSGS RCSVSAVNLP KHVDSIINKR LSKSSATLWN SPSRNRSLQL SAWESSIVDR LMTPTLSFLA RSRSAVTLPR NGRDQGRGCD PGRGPTWGRA GASLARGPQP DRTHPSAAVP VCPRSASASP LTPCSVTRSV HRCAPAGERG ERRKPNAGGS PAPVRRRPEA SPVQKKEKKD KERENEKEKS ALARERSLKK RQSLPASPRA RLSASTASEL S; Positive Samples: LO2, Jurkat, HeLa, Mouse liver; Cellular Location: Cytoplasm, cytoskeleton, spindle.

MAP7D1 (MAP7 Domain Containing 1) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MAP7D1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Map7d1
Gene Accession No. A2AJI0, Q3KQU3
Gene Alias arginine/proline rich coiled-coil 1; arginine/proline-rich coiled-coil domain-containing protein 1; AV028413; BC019977; FLJ10350; FLJ39022; KIAA1187; MAP7 domain containing 1; MAP7 domain-containing protein 1; MAP7D1; microtubule-associated protein 7 domain containing 1; mKIAA1187; Mtap7d1; Parcc1; PP2464; proline arginine rich coiled coil 1; proline/arginine-rich coiled-coil domain-containing protein 1; Rprc1
Gene Symbols Map7d1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-460 of human MAP7D1 (NP_0605373).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245877, 55700
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.