Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP7D1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988980
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8980 Supplier Bioss Supplier No. BS9314R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MAP7D1 (MAP7 Domain Containing 1) is a Protein Coding gene.

Specifications

Antigen MAP7D1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Map7d1
Gene Accession No. Q3KQU3
Gene Alias arginine/proline rich coiled-coil 1; arginine/proline rich coiled-coil 1-like; Arginine/proline-rich coiled-coil domain-containing protein 1; AV028413; BC019977; FLJ10350; FLJ39022; Kiaa1187; LOC100355828; MAP7 domain containing 1; MAP7 domain-containing protein 1; MAP7D1; microtubule-associated protein 7 domain containing 1; mKIAA1187; Mtap7d1; PARCC1; PP2464; proline arginine rich coiled coil 1; Proline/arginine-rich coiled-coil domain-containing protein 1; RPRC1
Gene Symbols Map7d1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MAP7D1/RPRC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55700
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.