Learn More
MAP7D3 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304935A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Promotes the assembly and stability of microtubules.Specifications
MAP7D3 | |
Polyclonal | |
Unconjugated | |
MAP7D3 | |
FLJ12649, MDP3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79649 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8IWC1 | |
MAP7D3 | |
Between 826 and 876 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.