MAPA™ Harpon Gloves

Manufacturer: MAPA™ 321127

 View more versions of this product

Catalog No. 19-130-2642

  Check Availability
Add to Cart

    Specifications

    Certifications CE
    Glove Cuff Style Pinked
    Hem Color Pink
    For Use With (Application) fishing, fish handling, oyster farming, work with frozen products
    Length (English) 12.5 in.
    Latex Free No
    Length (Metric) 31.7cm
    Material Nitrile
    Size 7
    Thickness 50mil
    Coating Material Rough
    Color Orange
    Lining Knit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit