Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPA™ Cobra™ FluoroSolv™ Gloves
Click to view available options
Size:
8
9
10
11
Length (English):
11 in.
14 in.
Length (Metric):
28 cm
35.5 cm
Description
Unsupported powder-free fluoroelastomer/nitrile gloves resist aromatic solvents, chloroform, and PCBs. Suitable for use in refineries, plating operations, laboratories, and hazardous material handling.
- Consist of a layer of black FluoroSolv fluoroelastomer polymer over a green, powder-free nitrile shell
- Two-tone layering serves as a visual wear indicator
- Nitrile layer also provides increased cut, puncture, and tear resistance
- Z-pattern texture channels chemicals away from glove surface, improving wet and dry grip
- Hand-specific and unlined with a flat cuff
- Embossed with size measurement for easy identification
- Style F-466 is 11 in. long (28 cm), 15 mil thick
- Style F-467 is 14 in. long (36 cm), 25 mil thick
Not for use with ketonic solvents.
Specifications
Specifications
Powdered | Powder Free |
Size | 11 |
Length (English) | 14 in. |
Length (Metric) | 35.5 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction