Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ Krynit Grip and Proof 580 Cut-Resistant Gloves, Nitrile Coated
GREENER_CHOICE

Catalog No. p-4912092 Shop All MAPA Professional Products
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Grip and proof nitrile coating on the palm and fingers ensures a secure grip in oily environments. Oil-proof gloves are ideal for jobs such as handling sheet metal, working with oily mechanical parts or sorting small, sharp parts.

  • Nitrile coating on the palm and fingers ensures secure grip in oily environments
  • Ideal for handling sheet metal, working with oily mechanical parts and sorting small and sharp parts
  • HDPE provides cut and abrasion resistance
  • CE level 3 cut resistance/ANSI A2
  • Medium weight

Specifications

Certifications/Compliance (Europe) CE
Coating Material Nitrile
Color Black
Cut Resistance Rating ANSI A2
Material Nitrile
Product Type Cut-resistant Gloves
Size 7
For Use With (Application) Handling sheet metal, working with oily mechanical parts, sorting small/sharp parts
Glove Cuff Style Knit
Model 580417
Series Krynit Grip & Proof 580
Length (English) 9 to 10 in.
Length (Metric) 22.8 to 25.4 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.