Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ Chem-Ply™ N-440 Chemical Protection Polychloroprene Gloves

Catalog No. 189992901 Shop All MAPA Professional Products
Click to view available options
Size:
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Medium-weight protection from a broad range of chemicals. Red inner layer ensures that any wear is clearly visible. Ideal for chemical and mechanical industries.

  • Resistant to numerous solvents, oils, acids, alcohols and alkalis
  • Straight cuff design
  • Embossed pattern finish ensures secure grip
  • Chlorinated interior for easy donning and doffing
  • 14 in. length
  • 30 mil thickness

Specifications

Certifications/Compliance NFPA 1992-1994
Certifications/Compliance (Europe) Cat 3, CE 0334
Chlorinated Yes
Color Black, Red
Latex Free Yes
Material Neoprene
Size 11
Thickness (English) 30 mil
Thickness (Metric) 0.76 mm
EN388 Standard EN 388:2111X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ABCJLMNS
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Handling and transporting drums of chemical products, handling on alkylation units, site pollution control, etching/degreasing, surface treatment of metals
Glove Cuff Style Straight
Grip Design Embossed
Lining Unlined
Model 407951
Series Chem-Ply™ N-440
Length (English) 14 in.
Length (Metric) 35.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.