Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPA™ UltraNeo™ 450 Chemical Protection Neoprene/Natural Rubber Gloves
Shop All MAPA Professional Products
Click to view available options
Size:
6
7
8
9
10
11
Description
- Extra long cuff for better forearm protection
- Freedom of movement, due to the supple quality of the neoprene
- Comfortable cotton flock lining
- Embossed texture provides secure grip
- Ideal for chemical handling, loading and unloading
- Other applications include: Automotive/mechanical industry, spreading fertilizer, preparation of industrial glues, and industrial cleaning/maintenance
Specifications
Specifications
Certifications/Compliance (Europe) | Cat 3 CE 0334, EN 420 |
Color | Black |
Latex Free | No |
Material | Neoprene, Latex |
Size | 8 |
Thickness (English) | 30 mil |
Thickness (Metric) | 0.76 mm |
EN388 Standard | EN 388:2121X |
EN ISO 374-1 | Type A |
EN ISO 374-1 CHEMICAL LETTER CODES | ALMNST |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction