Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ TempDex 720 Nitrile-Coated Thermal Protection Gloves

Catalog No. 19046553 Shop All MAPA Professional Products
Click to view available options
Size:
7
9
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Aramid Fiber knit construction provides cut resistance, while also offering protection from temperatures up to 150°C.

  • Aramid Fiber knit construction offers cut protection
  • Terrycloth liner provides comfort and insulation
  • Nitrile coating enhances grip in moderately oily applications
  • Silicone free to prevent traces or defects on metal sheets and glass
  • Protection from temperatures up to 150°C

Specifications

Coating Material Nitrile
Color Orange
Material Aramid Fiber
Size 7
Temperature (English) Max. 300°F
Temperature (Metric) Max 150°C
For Use With (Application) Ceramic, Hot Material Handling, Steel Manufacturing, and Vulcanization
Glove Cuff Style Knit
Series TempDex 720
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.