Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Human MAPK1 (ERK2), GST Tag Recombinant Protein

Catalog No. p-7076066
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 μg

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots; aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length

MAPK1 (ERK2) is a serine/threonine kinase that acts as an important activator of p90 RSK, MSK, ELK1, and Stat3, and is involved in the MAPK pathway. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P28482
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.05% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 5594
Molecular Weight (g/mol) 69.7 kDa
Name Human MAPK1 (ERK2), GST Tag
Quantity 1 mg
Gene Alias 9030612K14Rik; AA407128; AU018647; C78273; ERK; ERK2; ERK-2; ERT1; extracellular signal-regulated kinase 2; extracellular signal-regulated kinase-2; extracellular-signal-regulated kinase 2; MAP kinase 1; MAP kinase 2; MAP kinase isoform p42; Mapk; MAPK 1; MAPK 2; Mapk1; MAPK2; mitogen activated protein kinase 1; mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; p38; p40; p41; p41mapk; P42MAPK; p42-MAPK; Prkm1; PRKM2; protein tyrosine kinase ERK2
Common Name ERK2
KinaseFamily MAP Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: CMGC Group
Shipping Condition Dry Ice
Expression System E. coli
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.