Learn More
MAPK1 Mouse anti-Canine, Human, Rat, Clone: OTI4D10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500469

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ERK2 (also known as MAPK1) is a member of the MAP kinase subfamily of Ser/Thr protein kinases. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets.Specifications
MAPK1 | |
Monoclonal | |
Unconjugated | |
MAPK1 | |
ERK, ERK-2, ERK2, ERT1, MAPK2, P42MAPK, PRKM1, PRKM2, p38, p40, p41, p41mapk, p42-MAPK | |
Mouse | |
Affinity Chromatography | |
RUO | |
116590, 477575, 5594 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
OTI4D10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P28482, P63086 | |
MAPK1 | |
Full length human recombit protein of human MAPK1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.