Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPK11, Rabbit anti-Human, Clone: E13-Q, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB15919
Specifications
MAPK11 | |
Monoclonal | |
Unconjugated | |
In 20 mM Tris-HCl buffer, pH 8.0 (10 mg/mL BSA, 0.05% Sodium Azide). | |
P38B/P38BETA2/PRKM11/SAPK2/SAPK2B/p38-2/p38Beta | |
Rabbit | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
E13-Q | |
ELISA (1:10000-1:20000) Immunocytochemistry Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
mitogen-activated protein kinase 11 | |
MAPK11 | |
A synthetic peptide corresponding to amino acids 352-363 at C-terminus of human MAPK11. | |
RUO | |
5600 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction