Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPK11, Rabbit anti-Human, FITC, Clone: P12-E, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB16011
Specifications
MAPK11 | |
Monoclonal | |
FITC | |
In PBS (10 mg/mL BSA, 0.05% Sodium Azide). | |
P38B/P38BETA2/PRKM11/SAPK2/SAPK2B/p38-2/p38Beta | |
Rabbit | |
RUO | |
5600 | |
Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
Liquid |
Flow Cytometry | |
P12-E | |
Flow Cytometry The optimal working dilution should be determined by the end user. | |
mitogen-activated protein kinase 11 | |
MAPK11 | |
100 Reactions | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction