Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK12 Chicken anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502207691
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7691 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7691 Supplier Fabgennix Inc Supplier No. MAPK12101Y
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

MAPK12 Polyclonal antibody specifically detects MAPK12 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen MAPK12
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Mapk12
Gene Accession No. O08911, P53778, Q63538
Gene Alias AW123708; ERK3; ERK5; erk6; ERK-6; etID309866.18; Extracellular signal-regulated kinase 6; fa05c12; LOW QUALITY PROTEIN: mitogen-activated protein kinase 12; mitogen-activated protein kinase 12; MAP kinase 12; MAP kinase p38 gamma; MAPK 12; mapk12; mapk12a; mitogen activated protein kinase 12; mitogen-activated protein kinase 12; mitogen-activated protein kinase 12a; mitogen-activated protein kinase 3; mitogen-activated protein kinase p38 gamma; OTTHUMP00000196671; P38gamma; p38-gamma; Prkm12; SAP kinase-3; sapk3; SAPK-3; stress activated protein kinase 3; stress-activated protein kinase 3; stress-activated protein kinase-3; wu:fa05c12; zgc:101695
Host Species Chicken
Immunogen Synthetic peptide taken within amino acid region 310-360 on human Mitogen-activated protein kinase 12.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29857, 60352, 6300
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.