Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MAPK12 (p38 gamma), His Tag Recombinant Protein

Catalog No. PV3654
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 μg

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

MAPK12 (p38 gamma) is a serine/threonine kinase involved in many different cellular processes. It is activated during hypoxia and gamma-irradiation-induced G(2) arrest. Activation of members of the mitogen-activated protein kinase family is a major mechanism for transduction of extracellular signals. Stress-activated protein kinases are one subclass of MAP kinases. MAPK12 functions as a signal transducer during differentiation of myoblasts to myotubes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P53778
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 6300
Molecular Weight (g/mol) 46 kDa
Name Human MAPK12 (p38 gamma), His Tag
Quantity 10 μg
Gene Alias AW123708; ERK3; ERK5; erk6; ERK-6; etID309866.18; Extracellular signal-regulated kinase 6; fa05c12; LOW QUALITY PROTEIN: mitogen-activated protein kinase 12; mitogen-activated protein kinase 12; MAP kinase 12; MAP kinase p38 gamma; MAPK 12; mapk12; mapk12a; mitogen activated protein kinase 12; mitogen-activated protein kinase 12; mitogen-activated protein kinase 12 A; mitogen-activated protein kinase 3; mitogen-activated protein kinase p38 gamma; OTTHUMP00000196671; P38gamma; p38-gamma; Prkm12; SAP kinase-3; sapk3; SAPK-3; stress activated protein kinase 3; stress-activated protein kinase 3; stress-activated protein kinase-3; wu:fa05c12; zgc:101695
Common Name p38 MAPK gamma
KinaseFamily MAP Kinase Family
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical terbium phospho-specific activity, Biochemical phospho-specific activity (no antibody)
Kinase Group Ser⁄Thr Kinases: CMGC Group
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™
Target Entry MAPK12
Target Gene mitogen-activated protein kinase 12
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.