Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPK12, Rabbit anti-Human, FITC, Clone: Q11-T, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB16012
Specifications
MAPK12 | |
Monoclonal | |
FITC | |
In PBS (10 mg/mL BSA, 0.05% Sodium Azide). | |
ERK3/ERK6/P38GAMMA/PRKM12/SAPK-3/SAPK3 | |
Rabbit | |
RUO | |
6300 | |
Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze. | |
Liquid |
Flow Cytometry | |
Q11-T | |
Flow Cytometry The optimal working dilution should be determined by the end user. | |
mitogen-activated protein kinase 12 | |
MAPK12 | |
100 Reactions | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction