Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAPK12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 201841AP150UL
Description
Activation of members of the mitogen-activated protein kinase family is a major mechanism for transduction of extracellular signals. Stress-activated protein kinases are one subclass of MAP kinases. The protein encoded by this gene functions as a signal transducer during differentiation of myoblasts to myotubes.Specifications
| MAPK12 | |
| Polyclonal | |
| Unconjugated | |
| MAPK12 | |
| ERK 6, ERK3, ERK6, MAP kinase 12, MAP kinase p38 gamma, MAPK 12, MAPK12, P38GAMMA, PRKM12, SAPK 3, SAPK3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29857, 60352, 6300 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08911, P53778, Q63538 | |
| Mapk12 | |
| MAPK12 Fusion Protein Ag14073 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction