Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK14 Mouse anti-Human, Unconjugated, Clone: CPTC-MAPK14-1, Abnova™

Catalog No. 89337654 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-654 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-654 Supplier Abnova Corporation Supplier No. MAB21161
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

MAPK14 Monoclonal antibody specifically detects MAPK14 in Human samples. It is validated for WB.

Specifications

Antigen MAPK14
Applications Western Blot
Classification Monoclonal
Clone CPTC-MAPK14-1
Conjugate Unconjugated
Description mitogen-activated protein kinase 14
Gene MAPK14
Gene Alias CSBP1, CSBP2, CSPB1, EXIP, Mxi2, PRKM14, PRKM15, RK, SAPK2A, p38, p38ALPHA
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human MAPK14
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1432
Target Species Human
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.