Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK14, Rabbit anti-Human, FITC, Clone: I15-E, Abnova™

Catalog No. 89998675 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-998-675 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-998-675 Supplier Abnova Corporation Supplier No. MAB16010
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen MAPK14
Applications Flow Cytometry
Classification Monoclonal
Clone I15-E
Conjugate FITC
Dilution Flow Cytometry The optimal working dilution should be determined by the end user.
Formulation In PBS (10 mg/mL BSA, 0.05% Sodium Azide).
Gene mitogen-activated protein kinase 14
Gene Alias CSBP1/CSBP2/CSPB1/EXIP/Mxi2/PRKM14/PRKM15/RK/SAPK2A/p38/p38ALPHA
Gene Symbols MAPK14
Host Species Rabbit
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1432
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.