Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK15, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89999240 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-240 Supplier Abnova Corporation Supplier No. PAB31665
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen MAPK15
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA (1:5000) Immunohistochemistry (1:100-300) Western Blot (1:500-2000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 0.5% BSA and 0.02% sodium azide).
Gene mitogen-activated protein kinase 15
Gene Alias ERK7/ERK8
Gene Symbols MAPK15
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 110-190 of human MAPK15 (phospho T175/Y177).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225689
Test Specificity This antibody detects endogenous levels of MAPK15 protein only when phosphorylated at T175/Y177.
Target Species Human, Mouse
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.