Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPK1IP1L Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571129
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1129 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-1129 Supplier Bethyl Laboratories, Inc Supplier No. A305743AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan

Mitogen-activated protein kinase 1-interacting protein 1-like (MAPK1IP1L) belongs to the MISS family.

Specifications

Antigen MAPK1IP1L
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene MAPK1IP1L
Gene Accession No. Q8BH93, Q8NDC0
Gene Alias C14orf32, MISS, c14_5346, MAPK-interacting and spindle-stabilizing protein, MAPK-interacting and spindle-stabilizing protein-like, mitogen activated protein kinase 1 interacting protein 1-like, mitogen-activated protein kinase 1-interacting protein 1-like
Gene Symbols MAPK1IP1L
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218975, 93487
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.